Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100129034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027406.1 | 1 | 3796 | |||
GenBank mRNA | AJ420421.1 | - | 1 | 41 | ||
BC042968.1 | - | 1 | 324 | |||
CR627469.1 | - | 1 | 1572 |
Gene Symbol: PSMB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056654.1 | - | 1 | 1419 |