Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAR2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046258.1 | 3 | 959 | |||
NR_046259.1 | 2 | 861 | ||||
GenBank mRNA | BC131487.1 | - | 1 | 1256 | ||
DA522902.1 | - | 3 | 188 | |||
DA866260.1 | - | 2 | 57 |
Gene Symbol: FAR2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026758.2 | 8 | 2511 | |||
GenBank mRNA | AK125921.1 | - | 8 | 2513 | ||
BC132694.1 | - | 3 | 209 | |||
BC132696.1 | - | 3 | 209 | |||
BC144438.1 | - | 3 | 209 | 9052961 | ||
EU668357.1 | - | 1 | 96 |