Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LINC00680-GUSBP4 SIRNA Assays ›
          Gene Symbol:
          LINC00680-GUSBP4 LINC00680
          Gene Name:
          LINC00680-GUSBP4 readthrough
          long intergenic non-protein coding RNA 680
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.6: 57919881 - 57961446 on Build GRCh38
          Chromosome Location:
          Chr.6: 57945834 - 57961446 on Build GRCh38
          UniGene ID:
          Hs.561539
          Hs.561539
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n269369

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LINC00680-GUSBP4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003660.2 2 166
          NR_132997.1 2 166
          GenBank mRNA BC144584.1 - 2 102
          BM553931.1 - 2 166
          DA269273.1 - 2 111
          Gene Symbol: LINC00680
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_125727.1 2 166
          NR_125728.1 2 166
          NR_125729.1 2 166
          NR_132994.1 2 166
          NR_132995.1 2 166
          GenBank mRNA AL541195.3 - 2 99
          BC011838.2 - 2 100
          BC017330.2 - 2 94
          BC030061.1 - 2 94
          BC065547.1 - 2 124
          BI756196.1 - 2 100
          DA613065.1 - 2 146
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI767251.1 - 2 101 Chr. 6 - 57947009 - 57961381 [-] on Build GRCh38
          GenBankmRNA BE294972.1 - 2 81 Chr. 6 - 57947051 - 57961361 [-] on Build GRCh38
          GenBankmRNA BG820556.1 - 2 97 Chr. 6 - 57946864 - 57961353 [-] on Build GRCh38
          GenBankmRNA BF303840.1 - 2 101 Chr. 6 - 57946901 - 57961380 [-] on Build GRCh38
          GenBankmRNA DA373197.1 - 2 69 Chr. 6 - 57947082 - 57961349 [-] on Build GRCh38
          GenBankmRNA BX363752.2 - 2 99 Chr. 6 - 57946765 - 57961379 [-] on Build GRCh38
          GenBankmRNA BI464906.1 - 118 Chr. - - [-] on Build
          GenBankmRNA DA485357.1 - 2 148 Chr. 6 - 57924570 - 57961428 [-] on Build GRCh38
          GenBankmRNA BM462641.1 - 102 Chr. - - [-] on Build
          GenBankmRNA CN278443.1 - 2 101 Chr. 6 - 57949889 - 57961380 [-] on Build GRCh38
          GenBankmRNA BE795382.1 - 2 80 Chr. 6 - 57946803 - 57961352 [-] on Build GRCh38
          GenBankmRNA BF205068.1 - 2 101 Chr. 6 - 57946986 - 57961380 [-] on Build GRCh38
          GenBankmRNA CB216393.1 - 2 84 Chr. 6 - 57947119 - 57961364 [-] on Build GRCh38
          GenBankmRNA BX473225.1 - 2 116 Chr. 6 - 57949047 - 57961395 [-] on Build GRCh38
          GenBankmRNA BI822492.1 - 2 74 Chr. 6 - 57946873 - 57961354 [-] on Build GRCh38
          GenBankmRNA BQ233003.1 - 2 123 Chr. 6 - 57949500 - 57961403 [-] on Build GRCh38
          GenBankmRNA DB276841.1 - 113 Chr. - - [-] on Build
          GenBankmRNA DA475413.1 - 2 148 Chr. 6 - 57948926 - 57961428 [-] on Build GRCh38
          GenBankmRNA BQ073163.1 - 2 115 Chr. 6 - 57946716 - 57961377 [-] on Build GRCh38
          GenBankmRNA DA385059.1 - 2 100 Chr. 6 - 57949659 - 57961380 [-] on Build GRCh38
          GenBankmRNA DA685652.1 - 2 146 Chr. 6 - 57949743 - 57961424 [-] on Build GRCh38
          GenBankmRNA DA445728.1 - 2 146 Chr. 6 - 57949686 - 57961424 [-] on Build GRCh38
          GenBankmRNA DA152646.1 - 2 145 Chr. 6 - 57949697 - 57961425 [-] on Build GRCh38
          GenBankmRNA BG682955.1 - 2 95 Chr. 6 - 57946784 - 57961374 [-] on Build GRCh38
          GenBankmRNA BM013198.1 - 2 93 Chr. 6 - 57946759 - 57961374 [-] on Build GRCh38
          GenBankmRNA DA053916.1 - 2 109 Chr. 6 - 57924337 - 57961389 [-] on Build GRCh38
          GenBankmRNA DA827265.1 - 2 146 Chr. 6 - 57948911 - 57961424 [-] on Build GRCh38
          GenBankmRNA DA485918.1 - 2 145 Chr. 6 - 57924546 - 57961425 [-] on Build GRCh38
          GenBankmRNA BI907421.1 - 2 87 Chr. 6 - 57924416 - 57961367 [-] on Build GRCh38
          GenBankmRNA DA429785.1 - 2 145 Chr. 6 - 57948934 - 57961425 [-] on Build GRCh38
          GenBankmRNA BF037692.1 - 1 51 Chr. 6 - 57946928 - 57961226 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix