Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DIABLO SIRNA Assays ›
          Gene Symbol:
          DIABLO
          Gene Name:
          diablo IAP-binding mitochondrial protein
          Gene Aliases:
          DFNA64, SMAC
          Chromosome Location:
          Chr.12: 122207662 - 122227534 on Build GRCh38
          UniGene ID:
          Hs.169611
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n270663

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DIABLO
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278302.1 NP_001265231.1 2 624
          NM_001278304.1 NP_001265233.1 1 1122
          NM_138930.3 NP_620308.1 2 624
          GenBank mRNA AK001399.1 - 1 1126
          BC095468.1 - 2 108 30557745
          BM746689.1 - 2 95
          CF128595.1 - 2 108
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CX787280.1 - 2 46 Chr. 12 - 122208285 - 122225990 [-] on Build GRCh38
          GenBankmRNA BU170405.1 - 1 28 Chr. 12 - 122208140 - 122224753 [-] on Build GRCh38
          GenBankmRNA BQ955339.1 - 91 Chr. - - [-] on Build
          GenBankmRNA CV813065.1 - 2 71 Chr. 12 - 122208293 - 122226015 [-] on Build GRCh38
          GenBankmRNA AA305624.1 - 2 85 Chr. 12 - 122216755 - 122226027 [-] on Build GRCh38
          GenBankmRNA BI194180.1 - 2 62 Chr. 12 - 122208383 - 122226005 [-] on Build GRCh38
          GenBankmRNA BI871724.1 - 1 321 Chr. 12 - 122208556 - 122225042 [-] on Build GRCh38
          GenBankmRNA CD106297.1 - 1 177 Chr. 12 - 122208307 - 122224905 [-] on Build GRCh38
          GenBankmRNA BG740235.1 - 2 64 Chr. 12 - 122208235 - 122226007 [-] on Build GRCh38
          GenBankmRNA AA156765.1 - 2 66 Chr. 12 - 122216502 - 122226012 [-] on Build GRCh38
          GenBankmRNA BM050184.1 - 98 Chr. - - [-] on Build
          GenBankmRNA HY052492.1 - 2 80 Chr. 12 - 122216557 - 122226024 [-] on Build GRCh38
          GenBankmRNA CN357804.1 - 2 69 Chr. 12 - 122208415 - 122226012 [-] on Build GRCh38
          GenBankmRNA BQ708206.1 - 2 78 Chr. 12 - 122208241 - 122226016 [-] on Build GRCh38
          GenBankmRNA BQ943434.1 - 2 78 Chr. 12 - 122208156 - 122226016 [-] on Build GRCh38
          GenBankmRNA HY141895.1 - 2 98 Chr. 12 - 122216755 - 122226041 [-] on Build GRCh38
          GenBankmRNA DB449949.1 - 2 106 Chr. 12 - 122216755 - 122226050 [-] on Build GRCh38
          GenBankmRNA DB140459.1 - 2 109 Chr. 12 - 122218309 - 122226053 [-] on Build GRCh38
          GenBankmRNA CV802708.1 - 2 96 Chr. 12 - 122216487 - 122226040 [-] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          extrinsic apoptotic signaling pathway via death domain receptors
          intrinsic apoptotic signaling pathway in response to oxidative stress
          activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
          positive regulation of apoptotic process
          neuron apoptotic process
          intrinsic apoptotic signaling pathway
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0