Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01011 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026856.1 | 3 | 1078 | |||
GenBank mRNA | AK055096.1 | - | 1 | 1975 | ||
AK123663.1 | - | 2 | 2313 | |||
BC108683.1 | - | 3 | 1293 | |||
BX640709.1 | - | 3 | 1076 |
Gene Symbol: LINC01410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121647.1 | 3 | 828 | |||
GenBank mRNA | BC039680.1 | - | 3 | 802 | ||
CR627148.1 | - | 3 | 829 |