Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other H19 SIRNA Assays ›
          Gene Symbol:
          H19 RPS12
          Gene Name:
          H19, imprinted maternally expressed transcript (non-protein coding)
          ribosomal protein S12
          Gene Aliases:
          ASM, ASM1, BWS, D11S813E, LINC00008, NCRNA00008, WT2
          S12
          Chromosome Location:
          Chr.11: 1995176 - 2001466 on Build GRCh38
          Chromosome Location:
          Chr.6: 132814569 - 132817564 on Build GRCh38
          UniGene ID:
          Hs.533566
          Hs.546289
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n272448

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: H19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002196.2 1 819
          NR_131223.1 1 819
          GenBank mRNA AK092486.1 - 2 270
          AL546946.3 - 1 776
          AL548405.3 - 1 778
          BC023213.1 - 1 131
          BC098439.1 - 1 179
          BC110657.1 - 1 418
          BC115700.1 - 1 462
          Gene Symbol: RPS12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC002550.1 - 609
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AL531884.3 - 1 88 Chr. 11 - 1995886 - 1997144 [-] on Build GRCh38
          GenBankmRNA AL545538.3 - 1 25 Chr. 11 - 1995647 - 1997081 [-] on Build GRCh38
          GenBankmRNA AL549829.3 - 1 142 Chr. 11 - 1995876 - 1997162 [-] on Build GRCh38
          GenBankmRNA AL550184.3 - 1 98 Chr. 11 - 1995747 - 1997154 [-] on Build GRCh38
          GenBankmRNA BX439190.2 - 1 97 Chr. 11 - 1995872 - 1997153 [-] on Build GRCh38
          GenBankmRNA BY794978.2 - 1 103 Chr. 11 - 1995179 - 1997159 [-] on Build GRCh38
          GenBankmRNA AL551585.3 - 11 Chr. 11 - 1995710 - 1997067 [-] on Build GRCh38
          GenBankmRNA AL546608.3 - 1 12 Chr. 11 - 1995870 - 1997068 [-] on Build GRCh38
          GenBankmRNA BX446459.2 - 1 70 Chr. 11 - 1995709 - 1997094 [-] on Build GRCh38
          GenBankmRNA M32053.3 - 1 1618 Chr. 11 - 1995181 - 1998676 [-] on Build GRCh38
          GenBankmRNA AL546892.3 - 1 90 Chr. 11 - 1996092 - 1997146 [-] on Build GRCh38
          GenBankmRNA AF125183.3 - 1 10885 Chr. 11|NT_187376.1 - 37387 - 63123 [-] on Build GRCh38
          GenBankmRNA AF087017.3 - 2 9113 Chr. 11 - 1965624 - 2006227 [-] on Build GRCh38
          GenBankmRNA BI091640.1 - 1 27 Chr. 11 - 1996099 - 1997083 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0