Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMEM198B SIRNA Assays ›
          Gene Symbol:
          TMEM198B
          Gene Name:
          transmembrane protein 198B (pseudogene)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 55829608 - 55836246 on Build GRCh38
          UniGene ID:
          Hs.731828
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n276636

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMEM198B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_036476.1 5 2098
          NR_036477.1 4 2005
          NR_036478.1 6 1754
          NR_036479.1 5 1610
          GenBank mRNA AF370398.1 - 2 776
          AK057179.1 - 4 926
          AK094261.1 - 1 779
          AK095340.1 - 5 1473
          BC072431.1 - 5 642
          BX648969.1 - 3 847
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BE672547.1 - 1 60 Chr. 12 - 55834876 - 55835392 [+] on Build GRCh38
          GenBankmRNA BI836674.1 - 5 580 Chr. 12 - 55830828 - 55835234 [+] on Build GRCh38
          GenBankmRNA BI768537.1 - 4 495 Chr. 12 - 55830858 - 55835272 [+] on Build GRCh38
          GenBankmRNA BF445153.1 - 1 31 Chr. 12 - 55834900 - 55835368 [+] on Build GRCh38
          GenBankmRNA BI753624.1 - 5 585 Chr. 12 - 55830848 - 55835196 [+] on Build GRCh38
          GenBankmRNA DA866465.1 - 5 525 Chr. 12 - 55830907 - 55834942 [+] on Build GRCh38
          GenBankmRNA BI768213.1 - 6 645 Chr. 12 - 55830880 - 55835188 [+] on Build GRCh38
          GenBankmRNA DT215055.1 - 4 449 Chr. 12 - 55830902 - 55835082 [+] on Build GRCh38
          GenBankmRNA BE348506.1 - 1 60 Chr. 12 - 55834876 - 55835425 [+] on Build GRCh38
          GenBankmRNA DB268252.1 - 5 551 Chr. 12 - 55830881 - 55834944 [+] on Build GRCh38
          GenBankmRNA DB035816.1 - 4 451 Chr. 12 - 55830901 - 55835050 [+] on Build GRCh38
          GenBankmRNA BQ880489.1 - 5 478 Chr. 12 - 55830930 - 55835235 [+] on Build GRCh38
          GenBankmRNA DA561263.1 - 5 516 Chr. 12 - 55830916 - 55834988 [+] on Build GRCh38
          GenBankmRNA BM919219.1 - 5 534 Chr. 12 - 55830893 - 55835439 [+] on Build GRCh38
          GenBankmRNA BI767471.1 - 502 Chr. - - [-] on Build
          GenBankmRNA BM543506.1 - 5 640 Chr. 12 - 55830859 - 55835199 [+] on Build GRCh38
          GenBankmRNA DA259378.1 - 5 547 Chr. 12 - 55830885 - 55834940 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0