Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other C17ORF62 SIRNA Assays ›
          Gene Symbol:
          C17orf62
          Gene Name:
          chromosome 17 open reading frame 62
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 82442586 - 82450831 on Build GRCh38
          UniGene ID:
          Hs.163113
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n284275

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: C17orf62
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_036519.1 2 253
          XM_006722293.2 XP_006722356.1 1 196
          XM_011523606.2 XP_011521908.1 1 196
          XM_017025072.1 XP_016880561.1 1 196
          XM_017025073.1 XP_016880562.1 1 196
          XM_017025075.1 XP_016880564.1 1 196
          GenBank mRNA DA436899.1 - 2 190
          DA453235.1 - 2 190
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA437330.1 - 1 577 Chr. 17 - 82450104 - 82450708 [-] on Build GRCh38
          GenBankmRNA BM544299.1 - 646 Chr. - - [-] on Build
          GenBankmRNA DA442919.1 - 2 183 Chr. 17 - 82449755 - 82450734 [-] on Build GRCh38
          GenBankmRNA DB162208.1 - 1 566 Chr. 17 - 82450103 - 82450699 [-] on Build GRCh38
          GenBankmRNA DA054719.1 - 1 449 Chr. 17 - 82450114 - 82450582 [-] on Build GRCh38
          GenBankmRNA DB110128.1 - 2 154 Chr. 17 - 82449717 - 82450732 [-] on Build GRCh38
          GenBankmRNA DA863090.1 - 1 576 Chr. 17 - 82450091 - 82450708 [-] on Build GRCh38
          GenBankmRNA DA444093.1 - 1 196 Chr. 17 - 82449754 - 82450329 [-] on Build GRCh38
          GenBankmRNA DA787680.1 - 2 159 Chr. 17 - 82449715 - 82450735 [-] on Build GRCh38
          GenBankmRNA DB291629.1 - 1 575 Chr. 17 - 82450121 - 82450708 [-] on Build GRCh38
          GenBankmRNA CR994172.1 - 1 541 Chr. 17 - 82449949 - 82450674 [-] on Build GRCh38
          GenBankmRNA CR612860.1 - 1 558 Chr. 17 - 82449070 - 82450691 [-] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0