Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC9 SIRNA Assays ›
          Gene Symbol:
          HDAC9
          Gene Name:
          histone deacetylase 9
          Gene Aliases:
          HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
          Chromosome Location:
          Chr.7: 18086942 - 19003745 on Build GRCh38
          UniGene ID:
          Hs.196054
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n290741

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HDAC9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321868.1 NP_001308797.1 26 9362
          NM_001321877.1 NP_001308806.1 25 9292
          NM_001321897.1 NP_001308826.1 26 9189
          NM_178423.2 NP_848510.1 26 9246
          NM_178425.3 NP_848512.1 25 9110
          XM_011515629.2 XP_011513931.1 26 9204
          XM_011515631.2 XP_011513933.1 27 9518
          XM_011515634.2 XP_011513936.1 25 9072
          XM_011515635.2 XP_011513937.1 27 10070
          XM_011515636.2 XP_011513938.1 26 9791
          XM_011515637.2 XP_011513939.1 26 9215
          XM_011515638.2 XP_011513940.1 26 9785
          XM_011515639.2 XP_011513941.1 26 9982
          XM_011515641.2 XP_011513943.1 27 10067
          XM_011515642.2 XP_011513944.1 25 10796
          XM_011515644.2 XP_011513946.1 27 10042
          XM_011515645.2 XP_011513947.1 25 10781
          XM_011515646.2 XP_011513948.1 25 10772
          XM_011515648.2 XP_011513950.1 26 9800
          XM_011515649.2 XP_011513951.1 26 10004
          XM_017012826.1 XP_016868315.1 26 9776
          XM_017012827.1 XP_016868316.1 27 9859
          XM_017012828.1 XP_016868317.1 25 9852
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AW953794.1 - 1 133 Chr. 7 - 19001789 - 19002416 [+] on Build GRCh38
          GenBankmRNA BX452228.2 - 1 293 Chr. 7 - 19001645 - 19002414 [+] on Build GRCh38
          GenBankmRNA CR622072.1 - 1 1078 Chr. 7 - 19000844 - 19002405 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          response to amphetamine
          transcription, DNA-templated
          inflammatory response
          heart development
          chromatin modification
          histone deacetylation
          neuron differentiation
          B cell differentiation
          cellular response to insulin stimulus
          peptidyl-lysine deacetylation
          B cell activation
          negative regulation of transcription, DNA-templated
          regulation of skeletal muscle fiber development
          regulation of striated muscle cell differentiation
          histone H3 deacetylation
          histone H4 deacetylation
          positive regulation of cell migration involved in sprouting angiogenesis
          transcription corepressor activity
          histone deacetylase activity
          protein kinase C binding
          protein binding
          transcription factor binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          protein deacetylase activity
          histone deacetylase binding
          metal ion binding
          repressing transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0