Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321961.1 | NP_001308890.1 | 11 | 1588 | ||
NM_001321962.1 | NP_001308891.1 | 10 | 1402 | |||
NM_001321963.1 | NP_001308892.1 | 12 | 2073 | |||
NM_001321964.1 | NP_001308893.1 | 12 | 2398 | |||
NM_001321965.1 | NP_001308894.1 | 13 | 2711 | |||
NM_001321966.1 | NP_001308895.1 | 11 | 1706 | |||
NM_012472.5 | NP_036604.2 | 12 | 1648 | |||
NR_073525.2 | 13 | 1748 | ||||
NR_135905.1 | 12 | 1737 | ||||
NR_135906.1 | 9 | 1178 | ||||
NR_135907.1 | 11 | 1424 | ||||
NR_135908.1 | 8 | 1118 | ||||
NR_135909.1 | 11 | 1542 | ||||
NR_135910.1 | 11 | 1849 | ||||
NR_135911.1 | 9 | 1928 | ||||
NR_135912.1 | 12 | 2487 | ||||
NR_135913.1 | 11 | 2174 | ||||
XM_006716538.3 | XP_006716601.2 | 12 | 1612 | |||
XM_011516950.2 | XP_011515252.1 | 11 | 1552 | |||
XM_017013296.1 | XP_016868785.1 | 11 | 2281 | |||
XM_017013297.1 | XP_016868786.1 | 12 | 1766 | |||
XM_017013298.1 | XP_016868787.1 | 14 | 1693 | |||
GenBank mRNA | BG204280.1 | - | 2 | 375 | ||
DA570320.1 | - | 1 | 19 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U60666.1 | - | 2356 | Chr. - - [-] on Build |