Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242894.1 | NP_001229823.1 | 7 | 4079 | ||
NM_001242895.1 | NP_001229824.1 | 5 | 3498 | |||
NM_024493.3 | NP_077819.2 | 6 | 3962 | |||
XM_005249423.3 | XP_005249480.1 | 5 | 4128 | |||
XM_006715215.2 | XP_006715278.1 | 6 | 3860 | |||
XM_006715216.2 | XP_006715279.1 | 6 | 3875 | |||
XM_006715218.3 | XP_006715281.1 | 5 | 3426 | |||
XM_017011320.1 | XP_016866809.1 | 7 | 3956 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR619760.1 | - | 2 | 378 | Chr. 6 - 28367205 - 28369155 [+] on Build GRCh38 |