Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251815.2 | XP_005251872.1 | 17 | 2104 | ||
XM_005251820.2 | XP_005251877.1 | 16 | 2050 | |||
XM_005251821.2 | XP_005251878.1 | 16 | 2047 | |||
XM_005251822.2 | XP_005251879.1 | 15 | 2035 | |||
XM_005251823.2 | XP_005251880.1 | 15 | 2032 | |||
XM_005251825.2 | XP_005251882.1 | 16 | 2017 | |||
XM_005251827.2 | XP_005251884.1 | 15 | 2005 | |||
XM_005251828.2 | XP_005251885.1 | 15 | 2002 | |||
XM_005251829.2 | XP_005251886.1 | 16 | 1960 | |||
XM_005251830.2 | XP_005251887.1 | 15 | 1948 | |||
XM_005251831.2 | XP_005251888.1 | 15 | 1867 | |||
XM_005251832.2 | XP_005251889.1 | 14 | 1852 | |||
XM_005251833.2 | XP_005251890.1 | 14 | 1849 | |||
XM_006717016.2 | XP_006717079.1 | 17 | 2047 | |||
XM_011518399.2 | XP_011516701.1 | 17 | 2134 | |||
XM_011518400.2 | XP_011516702.1 | 17 | 2131 | |||
XM_017014487.1 | XP_016869976.1 | 17 | 2044 | |||
XM_017014488.1 | XP_016869977.1 | 16 | 2020 | |||
XM_017014489.1 | XP_016869978.1 | 16 | 1963 | |||
XM_017014492.1 | XP_016869981.1 | 17 | 2420 | |||
XM_017014493.1 | XP_016869982.1 | 15 | 1864 | |||
GenBank mRNA | AK000975.1 | - | 14 | 1736 | ||
AK001616.1 | - | 14 | 1698 | |||
AK023681.1 | - | 16 | 1960 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC094851.1 | - | 1 | 807 | Chr. 9 - 129895800 - 129896635 [-] on Build GRCh38 |