Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MITF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198177.2 | NP_937820.1 | 1 | 21 | ||
XM_005264755.3 | XP_005264812.1 | 1 | 163 | |||
XM_011533723.2 | XP_011532025.1 | 1 | 165 | |||
GenBank mRNA | AB006989.1 | - | 11 | |||
DC388606.1 | - | 1 | 21 |