Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005249807.4 | XP_005249864.1 | 7 | 5390 | ||
XM_006715752.2 | XP_006715815.1 | 8 | 5358 | |||
XM_017012414.1 | XP_016867903.1 | 7 | 5317 | |||
XM_017012415.1 | XP_016867904.1 | 8 | 5419 | |||
XM_017012417.1 | XP_016867906.1 | 8 | 3821 | |||
XM_017012418.1 | XP_016867907.1 | 7 | 3691 | |||
XM_017012421.1 | XP_016867910.1 | 8 | 5541 | |||
XM_017012422.1 | XP_016867911.1 | 8 | 5430 | |||
XM_017012424.1 | XP_016867913.1 | 7 | 5333 | |||
XM_017012430.1 | XP_016867919.1 | 8 | 3793 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK024255.1 | - | 1 | 1813 | Chr. 7 - 43628315 - 43630620 [-] on Build GRCh38 |