Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIITA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522485.2 | XP_011520787.1 | 20 | 3831 |
Gene Symbol: LOC100505564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055529.1 | - | 2 | 416 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA559788.1 | - | 2 | 416 | Chr. 16 - 10940719 - 10941427 [+] on Build GRCh38 |