Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927759 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132750.1 | 1 | 2163 | |||
GenBank mRNA | AK097187.1 | - | 1 | 2163 |