Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SCAMP1-AS1 SIRNA Assays ›
          Gene Symbol:
          SCAMP1-AS1
          Gene Name:
          SCAMP1 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 78358764 - 78360541 on Build GRCh38
          UniGene ID:
          Hs.586193
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n329825

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SCAMP1-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_105014.1 2 594
          GenBank mRNA AK129849.1 - 1 673
          BC039455.1 - 2 399
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI753922.1 - 2 513 Chr. 5 - 78359366 - 78360461 [-] on Build GRCh38
          GenBankmRNA BI755224.1 - 2 514 Chr. 5 - 78359385 - 78360461 [-] on Build GRCh38
          GenBankmRNA BE883417.1 - 2 420 Chr. 5 - 78359566 - 78360368 [-] on Build GRCh38
          GenBankmRNA BQ420880.1 - 2 457 Chr. 5 - 78359430 - 78360404 [-] on Build GRCh38
          GenBankmRNA DB487477.1 - 2 398 Chr. 5 - 78359668 - 78360345 [-] on Build GRCh38
          GenBankmRNA BI667651.1 - 2 403 Chr. 5 - 78359483 - 78360347 [-] on Build GRCh38
          GenBankmRNA CN367798.1 - 2 430 Chr. 5 - 78359474 - 78360377 [-] on Build GRCh38
          GenBankmRNA AW135430.1 - 1 348 Chr. 5 - 78357435 - 78360067 [-] on Build GRCh38
          GenBankmRNA CB111278.1 - 2 448 Chr. 5 - 78359688 - 78360375 [-] on Build GRCh38
          GenBankmRNA BI260618.1 - 2 560 Chr. 5 - 78359523 - 78360507 [-] on Build GRCh38
          GenBankmRNA BX362462.2 - 2 442 Chr. 5 - 78359298 - 78360390 [-] on Build GRCh38
          GenBankmRNA BQ277495.1 - 2 428 Chr. 5 - 78359393 - 78360372 [-] on Build GRCh38
          GenBankmRNA BI755223.1 - 2 513 Chr. 5 - 78359395 - 78360461 [-] on Build GRCh38
          GenBankmRNA HY043796.1 - 2 411 Chr. 5 - 78359670 - 78360357 [-] on Build GRCh38
          GenBankmRNA BG911071.1 - 2 413 Chr. 5 - 78359263 - 78360359 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix