Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC015656.2 | - | 2 | 511 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG761124.1 | - | 2 | 512 | Chr. 10 - 98252023 - 98256273 [+] on Build GRCh38 |