Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720191.2 | XP_006720254.1 | 10 | 13984 | ||
XM_017021427.1 | XP_016876916.1 | 11 | 14226 | |||
XM_017021428.1 | XP_016876917.1 | 11 | 14211 |