Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERVK-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FN806845.1 | - | 1 | 473 |
Gene Symbol: KDM5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005262560.2 | XP_005262617.1 | 26 | 10842 | ||
XM_005262561.2 | XP_005262618.1 | 27 | 10746 | |||
XM_011531468.2 | XP_011529770.1 | 27 | 10899 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK096805.1 | - | 1 | 1210 | Chr. 20 - 35262214 - 35264187 [-] on Build GRCh38 |