Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GBAP1 SIRNA Assays ›
          Gene Symbol:
          GBAP1
          Gene Name:
          glucosylceramidase beta pseudogene 1
          Gene Aliases:
          GBAP
          Chromosome Location:
          Chr.1: 155213825 - 155227534 on Build GRCh38
          UniGene ID:
          Hs.719930
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n368355

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GBAP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA DB192327.1 - 3 484
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA080524.1 - 2 412 Chr. 1 - 155218067 - 155219004 [-] on Build GRCh38
          GenBankmRNA DA233497.1 - 2 412 Chr. 1 - 155218062 - 155219004 [-] on Build GRCh38
          GenBankmRNA DA449111.1 - 3 458 Chr. 1 - 155218119 - 155228016 [-] on Build GRCh38
          GenBankmRNA DA317149.1 - 2 416 Chr. 1 - 155218096 - 155219008 [-] on Build GRCh38
          GenBankmRNA DA224838.1 - 2 400 Chr. 1 - 155218058 - 155218993 [-] on Build GRCh38
          GenBankmRNA DB253703.1 - 2 413 Chr. 1 - 155218152 - 155219005 [-] on Build GRCh38
          GenBankmRNA DA068957.1 - 2 424 Chr. 1 - 155218074 - 155219016 [-] on Build GRCh38
          GenBankmRNA DA345905.1 - 3 440 Chr. 1 - 155218075 - 155227469 [-] on Build GRCh38
          GenBankmRNA DA353838.1 - 2 403 Chr. 1 - 155218055 - 155218995 [-] on Build GRCh38
          GenBankmRNA DA397642.1 - 2 413 Chr. 1 - 155218046 - 155219005 [-] on Build GRCh38
          GenBankmRNA HY139750.1 - 2 430 Chr. 1 - 155218184 - 155219022 [-] on Build GRCh38
          GenBankmRNA DA219155.1 - 2 396 Chr. 1 - 155218049 - 155218988 [-] on Build GRCh38
          GenBankmRNA DA532484.1 - 2 419 Chr. 1 - 155218175 - 155219011 [-] on Build GRCh38
          GenBankmRNA DA164501.1 - 2 425 Chr. 1 - 155218084 - 155219017 [-] on Build GRCh38
          GenBankmRNA DA233555.1 - 2 430 Chr. 1 - 155218065 - 155219022 [-] on Build GRCh38
          GenBankmRNA BQ218826.1 - 3 507 Chr. 1 - 155217920 - 155227522 [-] on Build GRCh38
          GenBankmRNA DA188934.1 - 2 438 Chr. 1 - 155218120 - 155219028 [-] on Build GRCh38
          GenBankmRNA DA057140.1 - 2 405 Chr. 1 - 155218051 - 155218997 [-] on Build GRCh38
          GenBankmRNA DA224846.1 - 2 448 Chr. 1 - 155218136 - 155219040 [-] on Build GRCh38
          GenBankmRNA DA769889.1 - 3 399 Chr. 1 - 155218071 - 155227430 [-] on Build GRCh38
          GenBankmRNA DA219124.1 - 2 396 Chr. 1 - 155218051 - 155218988 [-] on Build GRCh38
          GenBankmRNA DA107001.1 - 3 505 Chr. 1 - 155218155 - 155227533 [-] on Build GRCh38
          GenBankmRNA DA179266.1 - 2 407 Chr. 1 - 155218169 - 155218999 [-] on Build GRCh38
          GenBankmRNA BI770380.1 - 3 349 Chr. 1 - 155217738 - 155244525 [-] on Build GRCh38
          GenBankmRNA DA217255.1 - 2 431 Chr. 1 - 155218108 - 155219021 [-] on Build GRCh38
          GenBankmRNA DA124268.1 - 2 413 Chr. 1 - 155213464 - 155219005 [-] on Build GRCh38
          GenBankmRNA DA236364.1 - 2 396 Chr. 1 - 155218083 - 155218988 [-] on Build GRCh38
          GenBankmRNA DA876012.1 - 2 430 Chr. 1 - 155218088 - 155219022 [-] on Build GRCh38
          GenBankmRNA DA798350.1 - 2 441 Chr. 1 - 155218108 - 155219033 [-] on Build GRCh38
          GenBankmRNA DA338971.1 - 2 396 Chr. 1 - 155218037 - 155218988 [-] on Build GRCh38
          GenBankmRNA DA525119.1 - 3 397 Chr. 1 - 155218063 - 155227428 [-] on Build GRCh38
          GenBankmRNA DA221157.1 - 2 413 Chr. 1 - 155218115 - 155219004 [-] on Build GRCh38
          GenBankmRNA DA096069.1 - 3 508 Chr. 1 - 155218177 - 155227539 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix