Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERVK-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC092439.1 | - | 4 | 850 |
Gene Symbol: KDM5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005262560.2 | XP_005262617.1 | 26 | 11171 | ||
XM_005262561.2 | XP_005262618.1 | 27 | 11075 | |||
XM_011531468.2 | XP_011529770.1 | 27 | 11228 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC036118.1 | - | 1 | 1246 | Chr. 8 - 144795090 - 144796772 [+] on Build GRCh38 |