Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 1700008K24RIK SIRNA Assays ›
          Gene Symbol:
          1700008K24Rik
          Gene Name:
          RIKEN cDNA 1700008K24 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 49112264 - 49113533 on Build GRCm38
          UniGene ID:
          Mm.46140
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n392921

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 1700008K24Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_038141.1 2 530
          NR_038142.1 3 433
          NR_038143.1 3 439
          NR_038144.1 2 386
          NR_038145.1 2 269
          GenBank mRNA AK005780.1 - 3 417
          CF104157.1 - 2 396
          CF982619.1 - 3 380
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BU961756.1 - 3 344 Chr. 17 - 49112273 - 49113436 [-] on Build GRCm38
          GenBankmRNA CA464976.1 - 2 528 Chr. 17 - 49112367 - 49113528 [-] on Build GRCm38
          GenBankmRNA CA465328.1 - 231 Chr. - - [-] on Build
          GenBankmRNA BU961335.1 - 2 239 Chr. 17 - 49112267 - 49113503 [-] on Build GRCm38
          GenBankmRNA AI615293.1 - 2 324 Chr. 17 - 49112644 - 49113471 [-] on Build GRCm38
          GenBankmRNA CF983402.1 - 2 438 Chr. 17 - 49112594 - 49113442 [-] on Build GRCm38
          GenBankmRNA CB272558.1 - 2 530 Chr. 17 - 49112654 - 49113533 [-] on Build GRCm38
          GenBankmRNA CA464085.1 - 2 313 Chr. 17 - 49112273 - 49113459 [-] on Build GRCm38
          GenBankmRNA CF198292.1 - 2 487 Chr. 17 - 49112605 - 49113491 [-] on Build GRCm38
          GenBankmRNA BF018461.1 - 3 355 Chr. 17 - 49112613 - 49113455 [-] on Build GRCm38
          GenBankmRNA BU938221.1 - 250 Chr. - - [-] on Build
          GenBankmRNA CA466118.1 - 2 496 Chr. 17 - 49112443 - 49113496 [-] on Build GRCm38
          GenBankmRNA CF983413.1 - 3 381 Chr. 17 - 49112515 - 49113481 [-] on Build GRCm38
          GenBankmRNA CF106086.1 - 2 344 Chr. 17 - 49112475 - 49113491 [-] on Build GRCm38
          GenBankmRNA BU939114.1 - 2 492 Chr. 17 - 49112404 - 49113492 [-] on Build GRCm38
          GenBankmRNA CB272932.1 - 2 346 Chr. 17 - 49112492 - 49113493 [-] on Build GRCm38
          GenBankmRNA CF105693.1 - 2 487 Chr. 17 - 49112592 - 49113491 [-] on Build GRCm38
          GenBankmRNA CN841515.1 - 2 345 Chr. 17 - 49112420 - 49113491 [-] on Build GRCm38
          GenBankmRNA CA463915.1 - 2 267 Chr. 17 - 49112289 - 49113528 [-] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0