Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK148950.1 | - | 1 | 1249 | ||
AK149008.1 | - | 1 | 1097 | |||
AK149239.1 | - | 1 | 1161 |
Gene Symbol: 9430041P20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AV162270.2 | - | 1 | 561 | ||
BB093294.2 | - | 1 | 204 |