Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 2810001A02RIK SIRNA Assays ›
          Gene Symbol:
          2810001A02Rik
          Gene Name:
          RIKEN cDNA 2810001A02 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          UniGene ID:
          Mm.439929
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n401366

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 2810001A02Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK012625.1 - 199
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CX234730.1 - 1 191 Chr. 15 - 86213786 - 86214360 [-] on Build GRCm38
          GenBankmRNA CK793247.1 - 1 184 Chr. 15 - 86213754 - 86214341 [-] on Build GRCm38
          GenBankmRNA AW989427.1 - 1 137 Chr. 15 - 86213815 - 86214306 [-] on Build GRCm38
          GenBankmRNA W41038.1 - 182 Chr. - - [-] on Build
          GenBankmRNA BG087067.2 - 1 194 Chr. 15 - 86213773 - 86214363 [-] on Build GRCm38
          GenBankmRNA AA434975.1 - 1 159 Chr. 15 - 86213889 - 86214329 [-] on Build GRCm38
          GenBankmRNA BG971627.1 - 1 194 Chr. 15 - 86213773 - 86214363 [-] on Build GRCm38
          GenBankmRNA AA097377.1 - 1 125 Chr. 15 - 86213859 - 86214295 [-] on Build GRCm38
          GenBankmRNA BM937289.1 - 1 112 Chr. 15 - 86213617 - 86214281 [-] on Build GRCm38
          GenBankmRNA CN539193.1 - 1 174 Chr. 15 - 86213534 - 86214343 [-] on Build GRCm38
          GenBankmRNA BF539835.1 - 1 179 Chr. 15 - 86213773 - 86214342 [-] on Build GRCm38
          GenBankmRNA CK032080.1 - 1 185 Chr. 15 - 86213630 - 86214354 [-] on Build GRCm38
          GenBankmRNA BX515995.1 - 1 68 Chr. 15 - 86213817 - 86214237 [-] on Build GRCm38
          GenBankmRNA AA763674.1 - 1 161 Chr. 15 - 86213843 - 86214333 [-] on Build GRCm38
          GenBankmRNA BE635053.1 - 1 126 Chr. 15 - 86213823 - 86214295 [-] on Build GRCm38
          GenBankmRNA BQ892173.1 - 1 190 Chr. 15 - 86213421 - 86214359 [-] on Build GRCm38
          GenBankmRNA BF585342.1 - 1 216 Chr. 15 - 86213827 - 86214385 [-] on Build GRCm38
          GenBankmRNA W82199.1 - 200 Chr. - - [-] on Build

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix