Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 2810408I11RIK SIRNA Assays ›
          Gene Symbol:
          2810408I11Rik
          Gene Name:
          RIKEN cDNA 2810408I11 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 64679869 - 64690659 on Build GRCm38
          UniGene ID:
          Mm.69044
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n408749

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 2810408I11Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_038009.1 2 409
          GenBank mRNA AK013046.1 - 2 410
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BY712101.1 - 2 408 Chr. 1 - 64679871 - 64690658 [-] on Build GRCm38
          GenBankmRNA AI046712.1 - 2 311 Chr. 1 - 64680083 - 64690550 [-] on Build GRCm38
          GenBankmRNA AA061317.1 - 2 352 Chr. 1 - 64680129 - 64690603 [-] on Build GRCm38
          GenBankmRNA W66710.1 - 2 371 Chr. 1 - 64680106 - 64690614 [-] on Build GRCm38
          GenBankmRNA AA153296.1 - 2 318 Chr. 1 - 64679979 - 64690571 [-] on Build GRCm38
          GenBankmRNA BY166560.1 - 2 379 Chr. 1 - 64680127 - 64690628 [-] on Build GRCm38
          GenBankmRNA AV475615.1 - 2 353 Chr. 1 - 64680049 - 64690598 [-] on Build GRCm38
          GenBankmRNA CA464200.1 - 2 349 Chr. 1 - 64679864 - 64690596 [-] on Build GRCm38
          GenBankmRNA AV455857.1 - 2 361 Chr. 1 - 64680023 - 64690611 [-] on Build GRCm38
          GenBankmRNA CN836304.1 - 2 325 Chr. 1 - 64679891 - 64690572 [-] on Build GRCm38
          GenBankmRNA AA914217.1 - 2 352 Chr. 1 - 64680100 - 64690604 [-] on Build GRCm38
          GenBankmRNA CN841970.1 - 414 Chr. - - [-] on Build
          GenBankmRNA BY166546.1 - 2 379 Chr. 1 - 64680127 - 64690628 [-] on Build GRCm38
          GenBankmRNA AA823892.1 - 2 358 Chr. 1 - 64679996 - 64690609 [-] on Build GRCm38
          GenBankmRNA AA684180.1 - 2 357 Chr. 1 - 64680007 - 64690604 [-] on Build GRCm38
          GenBankmRNA AA110120.1 - 2 329 Chr. 1 - 64679990 - 64690582 [-] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0