Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PBX1 SIRNA Assays ›
          Gene Symbol:
          Pbx1
          Gene Name:
          pre B cell leukemia homeobox 1
          Gene Aliases:
          2310056B04Rik, D230003C07Rik, Pbx-1
          Chromosome Location:
          Chr.1: 168119364 - 168432264 on Build GRCm38
          UniGene ID:
          Mm.43358
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n414646

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pbx1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK083616.1 - 1 1582
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BY467573.1 - 1 296 Chr. 1 - 168153551 - 168153929 [-] on Build GRCm38
          GenBankmRNA BB516776.1 - 1 216 Chr. 1 - 168153551 - 168153849 [-] on Build GRCm38
          GenBankmRNA CJ258330.1 - 1 356 Chr. 1 - 168153551 - 168153989 [-] on Build GRCm38
          GenBankmRNA BB441155.1 - 1 189 Chr. 1 - 168153551 - 168153823 [-] on Build GRCm38
          GenBankmRNA BB537829.1 - 1 198 Chr. 1 - 168153551 - 168153831 [-] on Build GRCm38
          GenBankmRNA BB193586.1 - 1 211 Chr. 1 - 168153551 - 168153844 [-] on Build GRCm38
          GenBankmRNA BB448915.1 - 1 240 Chr. 1 - 168153551 - 168153862 [-] on Build GRCm38
          GenBankmRNA BB317352.1 - 1 233 Chr. 1 - 168153551 - 168153866 [-] on Build GRCm38
          GenBankmRNA BB040980.1 - 1 233 Chr. 1 - 168153551 - 168153866 [-] on Build GRCm38
          GenBankmRNA CB248204.1 - 1 782 Chr. 1 - 168153590 - 168154417 [-] on Build GRCm38
          GenBankmRNA CJ258792.1 - 1 422 Chr. 1 - 168153551 - 168154055 [-] on Build GRCm38
          GenBankmRNA DV552784.1 - 1 349 Chr. 1 - 168153529 - 168153981 [-] on Build GRCm38
          GenBankmRNA BB350195.1 - 1 189 Chr. 1 - 168153551 - 168153822 [-] on Build GRCm38
          GenBankmRNA BB532528.1 - 1 251 Chr. 1 - 168153551 - 168153884 [-] on Build GRCm38
          GenBankmRNA BB392910.1 - 1 245 Chr. 1 - 168153551 - 168153878 [-] on Build GRCm38
          GenBankmRNA BB222550.1 - 1 184 Chr. 1 - 168153551 - 168153817 [-] on Build GRCm38
          GenBankmRNA BF466686.1 - 1 244 Chr. 1 - 168153580 - 168153877 [-] on Build GRCm38
          GenBankmRNA BB226954.1 - 1 202 Chr. 1 - 168153551 - 168153835 [-] on Build GRCm38
          GenBankmRNA BB398026.1 - 1 238 Chr. 1 - 168153538 - 168153858 [-] on Build GRCm38
          GenBankmRNA BB027467.1 - 1 202 Chr. 1 - 168153551 - 168153835 [-] on Build GRCm38
          GenBankmRNA BB502650.1 - 1 231 Chr. 1 - 168153551 - 168153862 [-] on Build GRCm38
          GenBankmRNA BB227653.1 - 1 185 Chr. 1 - 168153551 - 168153791 [-] on Build GRCm38
          GenBankmRNA AV015754.1 - 1 255 Chr. 1 - 168153551 - 168153886 [-] on Build GRCm38
          GenBankmRNA BB257321.1 - 1 234 Chr. 1 - 168153551 - 168153867 [-] on Build GRCm38
          GenBankmRNA BB491463.1 - 1 223 Chr. 1 - 168153551 - 168153856 [-] on Build GRCm38
          GenBankmRNA BG087478.2 - 1 390 Chr. 1 - 168153552 - 168154023 [-] on Build GRCm38
          GenBankmRNA BB439099.1 - 1 199 Chr. 1 - 168153551 - 168153832 [-] on Build GRCm38
          GenBankmRNA BB362644.1 - 1 192 Chr. 1 - 168153551 - 168153825 [-] on Build GRCm38
          GenBankmRNA BB363234.1 - 1 201 Chr. 1 - 168153535 - 168153834 [-] on Build GRCm38
          GenBankmRNA BB531810.1 - 1 241 Chr. 1 - 168153551 - 168153863 [-] on Build GRCm38
          GenBankmRNA BB446070.2 - 1 540 Chr. 1 - 168153527 - 168154173 [-] on Build GRCm38
          GenBankmRNA BE981671.1 - 1 294 Chr. 1 - 168153547 - 168153927 [-] on Build GRCm38
          GenBankmRNA BE634095.1 - 1 308 Chr. 1 - 168153548 - 168153941 [-] on Build GRCm38
          GenBankmRNA DV047611.1 - 367 Chr. - - [-] on Build
          GenBankmRNA BB271560.1 - 1 212 Chr. 1 - 168153551 - 168153845 [-] on Build GRCm38
          GenBankmRNA BB228846.1 - 1 221 Chr. 1 - 168153551 - 168153855 [-] on Build GRCm38
          GenBankmRNA BB387836.1 - 1 257 Chr. 1 - 168153551 - 168153891 [-] on Build GRCm38
          GenBankmRNA BB313659.1 - 1 238 Chr. 1 - 168153551 - 168153866 [-] on Build GRCm38
          GenBankmRNA CJ259288.1 - 1 369 Chr. 1 - 168153551 - 168154002 [-] on Build GRCm38
          GenBankmRNA BB116333.1 - 1 221 Chr. 1 - 168153551 - 168153853 [-] on Build GRCm38
          GenBankmRNA CN533958.1 - 1 288 Chr. 1 - 168153532 - 168153884 [-] on Build GRCm38

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: BC044624, AL049381.

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0