Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RNF11 SIRNA Assays ›
          Gene Symbol:
          Rnf11
          Gene Name:
          ring finger protein 11
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 109452473 - 109484723 on Build GRCm38
          UniGene ID:
          Mm.392580
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n420167

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rnf11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK003552.1 - 1 460
          AK151082.1 - 977
          AK153482.1 - 978
          AK184946.1 - 1 334
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BE689385.1 - 1 215 Chr. 4 - 109452582 - 109453068 [-] on Build GRCm38
          GenBankmRNA AA986225.1 - 1 52 Chr. 4 - 109452338 - 109452905 [-] on Build GRCm38
          GenBankmRNA AV032797.2 - 1 539 Chr. 4 - 109452392 - 109453392 [-] on Build GRCm38
          GenBankmRNA CJ206508.1 - 1 104 Chr. 4 - 109452558 - 109452957 [-] on Build GRCm38
          GenBankmRNA AV455824.1 - 1 134 Chr. 4 - 109452553 - 109452974 [-] on Build GRCm38
          GenBankmRNA AU078819.2 - 1 107 Chr. 4 - 109452388 - 109452961 [-] on Build GRCm38
          GenBankmRNA AA177477.1 - 1 85 Chr. 4 - 109452547 - 109452938 [-] on Build GRCm38
          GenBankmRNA CB318378.1 - 286 Chr. - - [-] on Build
          GenBankmRNA BY763419.1 - 1 385 Chr. 4 - 109452561 - 109453221 [-] on Build GRCm38
          GenBankmRNA BY654266.1 - 1 156 Chr. 4 - 109452559 - 109453007 [-] on Build GRCm38
          GenBankmRNA CB524635.1 - 1 160 Chr. 4 - 109452553 - 109453013 [-] on Build GRCm38
          GenBankmRNA BI081496.1 - 319 Chr. - - [-] on Build
          GenBankmRNA BI663962.1 - 1 127 Chr. 4 - 109452382 - 109452980 [-] on Build GRCm38
          GenBankmRNA BF385908.1 - 1 57 Chr. 4 - 109452230 - 109452899 [-] on Build GRCm38
          GenBankmRNA BY376047.1 - 1 138 Chr. 4 - 109452558 - 109452991 [-] on Build GRCm38
          GenBankmRNA BF139947.1 - 1 245 Chr. 4 - 109452556 - 109453098 [-] on Build GRCm38
          GenBankmRNA AV348046.2 - 1 290 Chr. 4 - 109452479 - 109453143 [-] on Build GRCm38
          GenBankmRNA BU556149.1 - 1 38 Chr. 4 - 109452372 - 109452889 [-] on Build GRCm38
          GenBankmRNA CJ212365.1 - 1 96 Chr. 4 - 109452554 - 109452949 [-] on Build GRCm38
          GenBankmRNA BY651212.1 - 1 119 Chr. 4 - 109452558 - 109452950 [-] on Build GRCm38
          GenBankmRNA BX521265.1 - 1 78 Chr. 4 - 109452378 - 109452917 [-] on Build GRCm38
          GenBankmRNA DV043104.1 - 303 Chr. - - [-] on Build
          GenBankmRNA BB789468.1 - 1 125 Chr. 4 - 109452558 - 109452977 [-] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0