Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A330076H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038438.1 | - | 691 | |||
AK048029.1 | - | 3358 | ||||
AK080655.2 | - | 694 | ||||
AK082028.1 | - | 942 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB269654.2 | - | 2 | 278 | Chr. 7 - 61930162 - 61945250 [-] on Build GRCm38 |