Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KLHL24 SIRNA Assays ›
          Gene Symbol:
          Klhl24
          Gene Name:
          kelch-like 24
          Gene Aliases:
          1110046J11Rik, 4930429H24Rik, 6530402O20Rik, C85082
          Chromosome Location:
          Chr.16: 20097457 - 20129226 on Build GRCm38
          UniGene ID:
          Mm.392450
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n423660

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Klhl24
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK004187.1 - 1 877
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BG971966.1 - 1 79 Chr. 16 - 20128905 - 20129221 [+] on Build GRCm38
          GenBankmRNA AA475074.1 - 1 312 Chr. 16 - 20128671 - 20129162 [+] on Build GRCm38
          GenBankmRNA AV114894.1 - 1 58 Chr. 16 - 20128925 - 20129213 [+] on Build GRCm38
          GenBankmRNA CJ260877.1 - 1 232 Chr. 16 - 20128751 - 20129216 [+] on Build GRCm38
          GenBankmRNA BE943978.1 - 1 223 Chr. 16 - 20128765 - 20129117 [+] on Build GRCm38
          GenBankmRNA AA462064.1 - 1 176 Chr. 16 - 20128807 - 20129206 [+] on Build GRCm38
          GenBankmRNA BE656316.1 - 1 235 Chr. 16 - 20128758 - 20129214 [+] on Build GRCm38
          GenBankmRNA AV252633.1 - 1 49 Chr. 16 - 20128935 - 20129216 [+] on Build GRCm38
          GenBankmRNA DV067986.1 - 262 Chr. - - [-] on Build
          GenBankmRNA CJ258034.1 - 1 467 Chr. 16 - 20128540 - 20129225 [+] on Build GRCm38
          GenBankmRNA CJ253123.1 - 1 185 Chr. 16 - 20128797 - 20129216 [+] on Build GRCm38
          GenBankmRNA AA273814.1 - 1 207 Chr. 16 - 20128776 - 20129215 [+] on Build GRCm38
          GenBankmRNA CJ258500.1 - 1 217 Chr. 16 - 20128766 - 20129225 [+] on Build GRCm38
          GenBankmRNA AA154835.1 - 1 291 Chr. 16 - 20128692 - 20129143 [+] on Build GRCm38
          GenBankmRNA BG092009.1 - 1 105 Chr. 16 - 20128878 - 20129209 [+] on Build GRCm38
          GenBankmRNA BY704431.1 - 1 877 Chr. 16 - 20128106 - 20129055 [+] on Build GRCm38
          GenBankmRNA BI104966.1 - 1 627 Chr. 16 - 20128353 - 20129106 [+] on Build GRCm38
          GenBankmRNA CJ261132.1 - 1 205 Chr. 16 - 20128778 - 20129216 [+] on Build GRCm38
          GenBankmRNA BB695752.1 - 1 207 Chr. 16 - 20128776 - 20129216 [+] on Build GRCm38
          GenBankmRNA BE336045.1 - 1 290 Chr. 16 - 20128693 - 20129114 [+] on Build GRCm38
          GenBankmRNA CJ260241.1 - 1 204 Chr. 16 - 20128780 - 20129225 [+] on Build GRCm38
          GenBankmRNA CJ263781.1 - 1 229 Chr. 16 - 20128754 - 20129216 [+] on Build GRCm38
          GenBankmRNA BI665692.1 - 1 357 Chr. 16 - 20128626 - 20129221 [+] on Build GRCm38
          GenBankmRNA BF151473.1 - 1 290 Chr. 16 - 20128693 - 20129190 [+] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-867f8cf58d-sfwzd:80/100.66.130.62:80.
          git-commit: 7a7e1ef32845d1da800a461bcf18a29ea3e3afc0
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix