Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139182.1 | - | 303 | |||
AK151082.1 | - | 1483 | ||||
AK153482.1 | - | 1484 |