Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other COL1A1 SIRNA Assays ›
          Gene Symbol:
          Col1a1
          Gene Name:
          collagen, type I, alpha 1
          Gene Aliases:
          Col1a-1, Cola-1, Cola1, Mov-13, Mov13
          Chromosome Location:
          Chr.11: 94936270 - 94951867 on Build GRCm38
          UniGene ID:
          Mm.277735
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n423864

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Col1a1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK159300.1 - 1248
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BY688164.1 - 1 207 Chr. 11 - 94952684 - 94953045 [+] on Build GRCm38
          GenBankmRNA AI157061.1 - 1 64 Chr. 11 - 94952827 - 94953032 [+] on Build GRCm38
          GenBankmRNA BY684701.1 - 1 239 Chr. 11 - 94952658 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY687118.1 - 1 250 Chr. 11 - 94952649 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY686391.1 - 1 178 Chr. 11 - 94952714 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY131999.1 - 1 259 Chr. 11 - 94952657 - 94952990 [+] on Build GRCm38
          GenBankmRNA BY686682.1 - 1 267 Chr. 11 - 94952622 - 94953040 [+] on Build GRCm38
          GenBankmRNA AA959755.1 - 1 216 Chr. 11 - 94952675 - 94953016 [+] on Build GRCm38
          GenBankmRNA BY685345.1 - 1 226 Chr. 11 - 94952666 - 94953037 [+] on Build GRCm38
          GenBankmRNA BY685941.1 - 1 223 Chr. 11 - 94952677 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY341330.1 - 1 231 Chr. 11 - 94952660 - 94952986 [+] on Build GRCm38
          GenBankmRNA AA049256.1 - 1 164 Chr. 11 - 94952747 - 94953037 [+] on Build GRCm38
          GenBankmRNA BY685712.1 - 1 228 Chr. 11 - 94952663 - 94953040 [+] on Build GRCm38
          GenBankmRNA W10457.1 - 159 Chr. - - [-] on Build
          GenBankmRNA AA562685.1 - 1 218 Chr. 11 - 94952673 - 94953037 [+] on Build GRCm38
          GenBankmRNA BY685716.1 - 1 223 Chr. 11 - 94952672 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY688246.1 - 2 266 Chr. 11 - 94951809 - 94953040 [+] on Build GRCm38
          GenBankmRNA AA645084.1 - 1 223 Chr. 11 - 94952668 - 94953036 [+] on Build GRCm38
          GenBankmRNA BY686486.1 - 1 280 Chr. 11 - 94952607 - 94953040 [+] on Build GRCm38
          GenBankmRNA BE136415.1 - 1 216 Chr. 11 - 94952675 - 94953022 [+] on Build GRCm38
          GenBankmRNA BY688405.1 - 1 220 Chr. 11 - 94952678 - 94953040 [+] on Build GRCm38
          GenBankmRNA CV304869.1 - 1 60 Chr. 11 - 94952835 - 94953042 [+] on Build GRCm38
          GenBankmRNA BY684148.1 - 1 225 Chr. 11 - 94952667 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY684975.1 - 1 215 Chr. 11 - 94952688 - 94953036 [+] on Build GRCm38
          GenBankmRNA BY598046.1 - 1 329 Chr. 11 - 94952554 - 94953031 [+] on Build GRCm38
          GenBankmRNA CJ177679.1 - 1 263 Chr. 11 - 94952655 - 94953042 [+] on Build GRCm38
          GenBankmRNA BY697059.1 - 1 285 Chr. 11 - 94952605 - 94953040 [+] on Build GRCm38
          GenBankmRNA AA051510.1 - 1 64 Chr. 11 - 94952827 - 94953021 [+] on Build GRCm38
          GenBankmRNA BY684550.1 - 1 205 Chr. 11 - 94952687 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY685118.1 - 1 229 Chr. 11 - 94952662 - 94953040 [+] on Build GRCm38
          GenBankmRNA CJ238482.1 - 1 543 Chr. 11 - 94952351 - 94953036 [+] on Build GRCm38
          GenBankmRNA BY683841.1 - 214 Chr. - - [-] on Build
          GenBankmRNA AA200783.1 - 1 120 Chr. 11 - 94952772 - 94953041 [+] on Build GRCm38
          GenBankmRNA BY685687.1 - 1 271 Chr. 11 - 94952617 - 94953036 [+] on Build GRCm38
          GenBankmRNA CB202290.1 - 488 Chr. - - [-] on Build
          GenBankmRNA AI646083.1 - 1 216 Chr. 11 - 94952675 - 94953015 [+] on Build GRCm38
          GenBankmRNA BY686329.1 - 1 270 Chr. 11 - 94952617 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY684706.1 - 1 228 Chr. 11 - 94952663 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY687113.1 - 1 241 Chr. 11 - 94952651 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY688497.1 - 1 248 Chr. 11 - 94952650 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY702242.1 - 1 217 Chr. 11 - 94952677 - 94953036 [+] on Build GRCm38
          GenBankmRNA BY684552.1 - 1 210 Chr. 11 - 94952682 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY684913.1 - 1 235 Chr. 11 - 94952657 - 94953036 [+] on Build GRCm38
          GenBankmRNA BY699387.1 - 1 235 Chr. 11 - 94952664 - 94953040 [+] on Build GRCm38
          GenBankmRNA BY686864.1 - 1 270 Chr. 11 - 94952617 - 94953036 [+] on Build GRCm38
          GenBankmRNA BY687466.1 - 1 241 Chr. 11 - 94952658 - 94953036 [+] on Build GRCm38

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: Z78279, CO397981.

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline