Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 2610528A11RIK SIRNA Assays ›
          Gene Symbol:
          2610528A11Rik
          Gene Name:
          RIKEN cDNA 2610528A11 gene
          Gene Aliases:
          AW494830
          Chromosome Location:
          Chr.14: 37102140 - 37110101 on Build GRCm38
          UniGene ID:
          Mm.271619
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n425618

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 2610528A11Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK012157.1 - 3 250
          BC049685.1 - 196
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BF580962.1 - 3 231 Chr. 14 - 37102167 - 37110085 [-] on Build GRCm38
          GenBankmRNA BU522916.1 - 3 234 Chr. 14 - 37102096 - 37110088 [-] on Build GRCm38
          GenBankmRNA BF582268.1 - 180 Chr. - - [-] on Build
          GenBankmRNA BQ939171.1 - 3 219 Chr. 14 - 37102148 - 37110064 [-] on Build GRCm38
          GenBankmRNA BY007317.1 - 3 247 Chr. 14 - 37102440 - 37110100 [-] on Build GRCm38
          GenBankmRNA AA469817.1 - 3 244 Chr. 14 - 37102449 - 37110087 [-] on Build GRCm38
          GenBankmRNA AA538362.1 - 3 220 Chr. 14 - 37102314 - 37110062 [-] on Build GRCm38
          GenBankmRNA AA272697.1 - 3 234 Chr. 14 - 37102361 - 37110087 [-] on Build GRCm38
          GenBankmRNA BX524139.1 - 3 234 Chr. 14 - 37102352 - 37110085 [-] on Build GRCm38
          GenBankmRNA AA463145.1 - 3 188 Chr. 14 - 37102307 - 37110028 [-] on Build GRCm38
          GenBankmRNA BY711346.1 - 245 Chr. - - [-] on Build
          GenBankmRNA W08316.1 - 235 Chr. - - [-] on Build
          GenBankmRNA DV552390.1 - 3 227 Chr. 14 - 37102502 - 37110081 [-] on Build GRCm38
          GenBankmRNA BG966710.1 - 3 233 Chr. 14 - 37102161 - 37110087 [-] on Build GRCm38
          GenBankmRNA BU704690.1 - 3 157 Chr. 14 - 37102296 - 37110011 [-] on Build GRCm38
          GenBankmRNA AA272358.1 - 3 230 Chr. 14 - 37102355 - 37110084 [-] on Build GRCm38
          GenBankmRNA AA238890.1 - 3 243 Chr. 14 - 37102328 - 37110096 [-] on Build GRCm38
          GenBankmRNA AA272330.1 - 3 230 Chr. 14 - 37102438 - 37110084 [-] on Build GRCm38
          GenBankmRNA AW989615.1 - 3 231 Chr. 14 - 37102429 - 37110085 [-] on Build GRCm38
          GenBankmRNA AA274151.1 - 3 243 Chr. 14 - 37102376 - 37110089 [-] on Build GRCm38
          GenBankmRNA AA509491.1 - 3 242 Chr. 14 - 37102624 - 37110089 [-] on Build GRCm38
          GenBankmRNA AA222093.1 - 3 242 Chr. 14 - 37102439 - 37110082 [-] on Build GRCm38
          GenBankmRNA AA265314.1 - 3 242 Chr. 14 - 37102444 - 37110096 [-] on Build GRCm38
          GenBankmRNA AA267641.1 - 3 247 Chr. 14 - 37102440 - 37110090 [-] on Build GRCm38
          GenBankmRNA AA790672.1 - 3 190 Chr. 14 - 37102609 - 37110045 [-] on Build GRCm38
          GenBankmRNA AA242158.1 - 3 245 Chr. 14 - 37102452 - 37110087 [-] on Build GRCm38
          GenBankmRNA AA017989.1 - 3 230 Chr. 14 - 37102417 - 37110084 [-] on Build GRCm38
          GenBankmRNA AA220337.1 - 3 243 Chr. 14 - 37102342 - 37110096 [-] on Build GRCm38
          GenBankmRNA AA538434.1 - 3 241 Chr. 14 - 37102346 - 37110087 [-] on Build GRCm38
          GenBankmRNA AA265120.1 - 3 230 Chr. 14 - 37102537 - 37110071 [-] on Build GRCm38
          GenBankmRNA AA529197.1 - 3 246 Chr. 14 - 37102439 - 37110087 [-] on Build GRCm38
          GenBankmRNA AA528993.1 - 3 170 Chr. 14 - 37102373 - 37110024 [-] on Build GRCm38
          GenBankmRNA BG968801.1 - 3 233 Chr. 14 - 37102156 - 37110082 [-] on Build GRCm38
          GenBankmRNA AA230541.1 - 3 244 Chr. 14 - 37102375 - 37110088 [-] on Build GRCm38
          GenBankmRNA AA498458.1 - 3 246 Chr. 14 - 37102439 - 37110087 [-] on Build GRCm38
          GenBankmRNA AA265055.1 - 3 243 Chr. 14 - 37102376 - 37110096 [-] on Build GRCm38
          GenBankmRNA AA240277.1 - 3 226 Chr. 14 - 37102376 - 37110071 [-] on Build GRCm38
          GenBankmRNA AA840147.1 - 3 206 Chr. 14 - 37102420 - 37110060 [-] on Build GRCm38
          GenBankmRNA BF531305.1 - 3 232 Chr. 14 - 37102143 - 37110086 [-] on Build GRCm38
          GenBankmRNA BB864020.1 - 3 250 Chr. 14 - 37102448 - 37110101 [-] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix