Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3110039I08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014147.1 | - | 476 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY713125.1 | - | 475 | Chr. - - [-] on Build |