Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120630.1 | 3 | 685 | |||
NR_120631.1 | 2 | 545 | ||||
GenBank mRNA | BC041007.2 | - | 2 | 545 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI457689.1 | - | 2 | 550 | Chr. 10 - 124704028 - 124713919 [+] on Build GRCh38 |