Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125431.1 | 2, 3 | 295 | |||
GenBank mRNA | CB992382.1 | - | 2, 3 | 300 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | R24121.1 | - | 255 | Chr. - - [-] on Build |