Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other THUMPD3-AS1 SIRNA Assays ›
          Gene Symbol:
          THUMPD3-AS1
          Gene Name:
          THUMPD3 antisense RNA 1
          Gene Aliases:
          SETD5-AS1
          Chromosome Location:
          Chr.3: 9385264 - 9397494 on Build GRCh38
          UniGene ID:
          Hs.598958
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n503738

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: THUMPD3-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027007.3 3 317
          NR_132780.1 4 391
          NR_132781.1 2 162
          GenBank mRNA AK093640.1 - 2 1442
          AL110136.1 - 3 313
          AW237468.1 - 2 162
          BC036698.1 - 2 2767
          BC044625.2 - 1 1428
          BC052961.1 - 3 2689
          BX953112.1 - 4 372
          HY142091.1 - 3 246
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BE217739.1 - 2 144 Chr. 3 - 9389511 - 9396605 [-] on Build GRCh38
          GenBankmRNA BJ994980.1 - 4 336 Chr. 3 - 9390267 - 9397439 [-] on Build GRCh38
          GenBankmRNA AA463541.1 - 3 311 Chr. 3 - 9390388 - 9397483 [-] on Build GRCh38
          GenBankmRNA BU159574.1 - 3 296 Chr. 3 - 9389825 - 9397464 [-] on Build GRCh38
          GenBankmRNA BM547994.1 - 1 370 Chr. 3 - 9389986 - 9390750 [-] on Build GRCh38
          GenBankmRNA DB239124.1 - 4 337 Chr. 3 - 9389699 - 9397439 [-] on Build GRCh38
          GenBankmRNA DB087998.1 - 3 263 Chr. 3 - 9389610 - 9397439 [-] on Build GRCh38
          GenBankmRNA CN349972.1 - 3 287 Chr. 3 - 9385793 - 9395580 [-] on Build GRCh38
          GenBankmRNA AA442499.1 - 2 229 Chr. 3 - 9390241 - 9391810 [-] on Build GRCh38
          GenBankmRNA CN349971.1 - 2 253 Chr. 3 - 9389492 - 9391828 [-] on Build GRCh38
          GenBankmRNA AA923144.1 - 3 300 Chr. 3 - 9389739 - 9397460 [-] on Build GRCh38
          GenBankmRNA BU537437.1 - 3 300 Chr. 3 - 9390067 - 9397474 [-] on Build GRCh38
          GenBankmRNA BX344859.2 - 3 289 Chr. 3 - 9389799 - 9397482 [-] on Build GRCh38
          GenBankmRNA BG751442.1 - 2 503 Chr. 3 - 9385793 - 9391828 [-] on Build GRCh38
          GenBankmRNA BQ428680.1 - 3 290 Chr. 3 - 9390025 - 9397496 [-] on Build GRCh38
          GenBankmRNA BI495272.1 - 1 466 Chr. 3 - 9390262 - 9390857 [-] on Build GRCh38
          GenBankmRNA AL524186.3 - 3 289 Chr. 3 - 9389625 - 9397482 [-] on Build GRCh38
          GenBankmRNA BI756773.1 - 2 265 Chr. 3 - 9389851 - 9391827 [-] on Build GRCh38
          GenBankmRNA DB523053.1 - 1 75 Chr. 3 - 9390015 - 9390474 [-] on Build GRCh38
          GenBankmRNA AI298618.1 - 3 304 Chr. 3 - 9390288 - 9396605 [-] on Build GRCh38
          GenBankmRNA BI830083.1 - 245 Chr. - - [-] on Build
          GenBankmRNA BQ548851.1 - 3 328 Chr. 3 - 9390138 - 9396634 [-] on Build GRCh38
          GenBankmRNA AI458342.1 - 3 279 Chr. 3 - 9389727 - 9396580 [-] on Build GRCh38
          GenBankmRNA BG943385.1 - 3 305 Chr. 3 - 9385854 - 9398579 [-] on Build GRCh38
          GenBankmRNA AA504762.1 - 3 320 Chr. 3 - 9390270 - 9395577 [-] on Build GRCh38
          GenBankmRNA DB497858.1 - 3 284 Chr. 3 - 9390186 - 9397462 [-] on Build GRCh38
          GenBankmRNA HY003086.1 - 3 319 Chr. 3 - 9390164 - 9397494 [-] on Build GRCh38
          GenBankmRNA BM712163.1 - 2 149 Chr. 3 - 9389556 - 9391729 [-] on Build GRCh38
          GenBankmRNA CR745754.1 - 2 219 Chr. 3 - 9390014 - 9391799 [-] on Build GRCh38
          GenBankmRNA CR743394.1 - 2 497 Chr. 3 - 9385898 - 9391823 [-] on Build GRCh38
          GenBankmRNA BX423431.2 - 2 493 Chr. 3 - 9389923 - 9391822 [-] on Build GRCh38
          GenBankmRNA HY159055.1 - 4 342 Chr. 3 - 9390261 - 9397439 [-] on Build GRCh38
          GenBankmRNA AW612073.1 - 3 294 Chr. 3 - 9390382 - 9396607 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0