Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108054.1 | 3 | 616 | |||
NR_108055.1 | 2 | 513 | ||||
NR_108056.1 | 2 | 386 | ||||
GenBank mRNA | AK095699.1 | - | 2 | 513 | ||
BX095585.1 | - | 2 | 386 | |||
DA500285.1 | - | 3 | 441 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA515194.1 | - | 2 | 513 | Chr. 10 - 4384246 - 4408772 [+] on Build GRCh38 |