Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ENTPD1-AS1 SIRNA Assays ›
          Gene Symbol:
          ENTPD1-AS1
          Gene Name:
          ENTPD1 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 95753206 - 96090235 on Build GRCh38
          UniGene ID:
          Hs.538374
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n505206

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ENTPD1-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_038444.1 2 286
          NR_134320.1 2 286
          NR_134321.1 2 286
          NR_134322.1 2 286
          GenBank mRNA AK091396.1 - 2 265
          AK096469.1 - 2 263
          AL120918.1 - 2 224
          BC028619.1 - 2 285
          BU852745.1 - 2 289
          DA419635.1 - 2 266
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BU854016.1 - 291 Chr. - - [-] on Build
          GenBankmRNA DB445857.1 - 2 272 Chr. 10 - 96057830 - 96090220 [-] on Build GRCh38
          GenBankmRNA BP871689.1 - 2 266 Chr. 10 - 96042608 - 96090215 [-] on Build GRCh38
          GenBankmRNA AV762020.1 - 2 263 Chr. 10 - 96055291 - 96090212 [-] on Build GRCh38
          GenBankmRNA BI225716.1 - 2 205 Chr. 10 - 95861313 - 96090155 [-] on Build GRCh38
          GenBankmRNA BI226415.1 - 2 205 Chr. 10 - 95861295 - 96090155 [-] on Build GRCh38
          GenBankmRNA BM549802.1 - 2 269 Chr. 10 - 95875320 - 96090202 [-] on Build GRCh38
          GenBankmRNA DA213255.1 - 2 266 Chr. 10 - 95876619 - 96090214 [-] on Build GRCh38
          GenBankmRNA AW732710.1 - 2 190 Chr. 10 - 95876567 - 96090139 [-] on Build GRCh38
          GenBankmRNA BE513929.1 - 2 186 Chr. 10 - 95861550 - 96090135 [-] on Build GRCh38
          GenBankmRNA BP871686.1 - 2 266 Chr. 10 - 96042601 - 96090215 [-] on Build GRCh38
          GenBankmRNA BE396593.1 - 2 186 Chr. 10 - 95861500 - 96090135 [-] on Build GRCh38
          GenBankmRNA CR739709.1 - 2 249 Chr. 10 - 96021352 - 96090196 [-] on Build GRCh38
          GenBankmRNA CD707141.1 - 2 308 Chr. 10 - 96089207 - 96090241 [-] on Build GRCh38
          GenBankmRNA DA491898.1 - 2 265 Chr. 10 - 95785184 - 96090214 [-] on Build GRCh38
          GenBankmRNA DA769118.1 - 2 263 Chr. 10 - 95921342 - 96090212 [-] on Build GRCh38
          GenBankmRNA BE562116.1 - 2 245 Chr. 10 - 95861292 - 96090150 [-] on Build GRCh38
          GenBankmRNA BE396777.1 - 2 185 Chr. 10 - 95861329 - 96090134 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0