Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027428.1 | 6 | 1358 | |||
NR_027429.1 | 5 | 1271 | ||||
NR_027430.1 | 5 | 1147 | ||||
NR_027431.2 | 5 | 1121 | ||||
NR_027432.1 | 4 | 1034 | ||||
GenBank mRNA | AK021673.1 | - | 5 | 990 | ||
AK096758.1 | - | 6 | 1060 | |||
AK308811.1 | - | 4 | 587 | |||
AK309654.1 | - | 5 | 770 | |||
AK310182.1 | - | 5 | 823 | |||
AK310261.1 | - | 4 | 724 | |||
BC028046.1 | - | 4 | 695 |