Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027428.1 | 6 | 1576 | |||
NR_027429.1 | 5 | 1489 | ||||
NR_027430.1 | 5 | 1365 | ||||
NR_027431.2 | 5 | 1339 | ||||
NR_027432.1 | 4 | 1252 | ||||
GenBank mRNA | AK021673.1 | - | 5 | 1208 | ||
AK096758.1 | - | 6 | 1278 | |||
AK308811.1 | - | 4 | 805 | |||
AK310261.1 | - | 4 | 942 | |||
BC028046.1 | - | 4 | 913 |