Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322800.1 | NP_001309729.1 | 2 | 1095 | ||
NR_136401.1 | 2 | 638 | ||||
NR_136402.1 | 2 | 270 | ||||
NR_136403.1 | 2 | 727 | ||||
GenBank mRNA | BM888151.1 | - | 2 | 259 |
Gene Symbol: CD68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW294871.1 | - | 1 | 72 |