Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110175.1 | 1 | 151 | |||
NR_110176.1 | 1 | 151 | ||||
NR_110177.1 | 1 | 151 | ||||
GenBank mRNA | CB047433.1 | - | 1 | 151 | ||
HY007950.1 | - | 1 | 151 |