Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015414.1 | 9 | 1031 | |||
NR_024276.1 | 5 | 545 | ||||
GenBank mRNA | BC039365.1 | - | 5 | 553 | ||
BC045586.1 | - | 9 | 1031 | |||
BC101990.1 | - | 3 | 421 | |||
BC101991.1 | - | 3 | 421 | |||
BC104745.1 | - | 5 | 625 | |||
BC104746.1 | - | 4 | 570 | |||
BC113056.1 | - | 3 | 421 | |||
BC113057.1 | - | 3 | 421 |