Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM163A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006711174.2 | XP_006711237.1 | 2 | 367 | ||
XM_006711175.2 | XP_006711238.1 | 2 | 357 | |||
XM_006711176.2 | XP_006711239.1 | 2 | 367 | |||
XM_006711177.1 | XP_006711240.1 | 2 | 199 | |||
XM_011509217.1 | XP_011507519.1 | 2 | 348 | |||
XM_011509221.1 | XP_011507523.1 | 2 | 185 | |||
XM_017000386.1 | XP_016855875.1 | 2 | 367 | |||
XM_017000387.1 | XP_016855876.1 | 2 | 367 | |||
XM_017000388.1 | XP_016855877.1 | 2 | 367 | |||
XM_017000389.1 | XP_016855878.1 | 2 | 367 | |||
XM_017000390.1 | XP_016855879.1 | 2 | 367 | |||
XM_017000391.1 | XP_016855880.1 | 2 | 367 | |||
GenBank mRNA | AK292432.1 | - | 2 | 183 | ||
AK302665.1 | - | 2 | 357 | |||
BC033359.1 | - | 2 | 317 |