Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGMB-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033932.1 | 2 | 568 | |||
GenBank mRNA | AK093265.1 | - | 3 | 399 | ||
BC042169.1 | - | 2 | 568 |