Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other THAP9-AS1 SIRNA Assays ›
          Gene Symbol:
          THAP9-AS1 SEC31A
          Gene Name:
          THAP9 antisense RNA 1
          SEC31 homolog A, COPII coat complex component
          Gene Aliases:
          -
          ABP125, ABP130, HSPC275, HSPC334, SEC31L1
          Chromosome Location:
          Chr.4: 82893452 - 82900916 on Build GRCh38
          Chromosome Location:
          Chr.4: 82818509 - 82900571 on Build GRCh38
          UniGene ID:
          Hs.728916
          Hs.370024
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n511208

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: THAP9-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_034075.1 6 935
          NR_034076.1 5 910
          NR_034077.1 4 836
          GenBank mRNA AF278605.1 - 1 353
          AK128047.1 - 1 3078
          BC020725.1 - 5 541
          BC045722.1 - 4 1297
          DA361777.1 - 4 460
          U92014.1 - 3 135
          Gene Symbol: SEC31A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA DA142666.1 - 4 482
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA833028.1 - 5 562 Chr. 4 - 82894668 - 82900566 [-] on Build GRCh38
          GenBankmRNA BU178413.1 - 4 396 Chr. 4 - 82894385 - 82900476 [-] on Build GRCh38
          GenBankmRNA BX339948.2 - 5 543 Chr. 4 - 82894370 - 82900549 [-] on Build GRCh38
          GenBankmRNA BE880640.1 - 4 424 Chr. 4 - 82894534 - 82900504 [-] on Build GRCh38
          GenBankmRNA T98067.1 - 135 Chr. - - [-] on Build
          GenBankmRNA DA925660.1 - 5 501 Chr. 4 - 82894740 - 82900506 [-] on Build GRCh38
          GenBankmRNA CB127731.1 - 5 501 Chr. 4 - 82894723 - 82900507 [-] on Build GRCh38
          GenBankmRNA DB476991.1 - 4 487 Chr. 4 - 82894795 - 82900566 [-] on Build GRCh38
          GenBankmRNA BU506989.1 - 5 468 Chr. 4 - 82894394 - 82900474 [-] on Build GRCh38
          GenBankmRNA BI762414.1 - 5 530 Chr. 4 - 82894631 - 82900536 [-] on Build GRCh38
          GenBankmRNA DA568200.1 - 5 541 Chr. 4 - 82894620 - 82900547 [-] on Build GRCh38
          GenBankmRNA BU175480.1 - 4 277 Chr. 4 - 82894208 - 82900173 [-] on Build GRCh38
          GenBankmRNA DA055649.1 - 4 489 Chr. 4 - 82894754 - 82900566 [-] on Build GRCh38
          GenBankmRNA DA615125.1 - 5 527 Chr. 4 - 82894781 - 82900536 [-] on Build GRCh38
          GenBankmRNA DA912133.1 - 4 428 Chr. 4 - 82894681 - 82900506 [-] on Build GRCh38
          GenBankmRNA BF886678.1 - 4 289 Chr. 4 - 82894665 - 82900159 [-] on Build GRCh38
          GenBankmRNA CN429666.1 - 5 483 Chr. 4 - 82894685 - 82900483 [-] on Build GRCh38
          GenBankmRNA BM854572.1 - 3 123 Chr. 4 - 82894455 - 82898005 [-] on Build GRCh38
          GenBankmRNA CN429665.1 - 4 381 Chr. 4 - 82894541 - 82900461 [-] on Build GRCh38
          GenBankmRNA AL603040.1 - 5 348 Chr. 4 - 82894613 - 82900354 [-] on Build GRCh38
          GenBankmRNA CN429663.1 - 5 416 Chr. 4 - 82894757 - 82900413 [-] on Build GRCh38
          GenBankmRNA BM478668.1 - 482 Chr. - - [-] on Build
          GenBankmRNA DA082618.1 - 4 488 Chr. 4 - 82894739 - 82900566 [-] on Build GRCh38
          GenBankmRNA BM472827.1 - 489 Chr. - - [-] on Build
          GenBankmRNA DA356680.1 - 4 432 Chr. 4 - 82894685 - 82900509 [-] on Build GRCh38
          GenBankmRNA R11369.1 - 3 136 Chr. 4 - 82894681 - 82898017 [-] on Build GRCh38
          GenBankmRNA DB030130.1 - 5 491 Chr. 4 - 82894751 - 82900496 [-] on Build GRCh38
          GenBankmRNA AL514590.3 - 5 452 Chr. 4 - 82894205 - 82900459 [-] on Build GRCh38
          GenBankmRNA DA958580.1 - 5 503 Chr. 4 - 82894765 - 82900506 [-] on Build GRCh38
          GenBankmRNA DB257486.1 - 5 530 Chr. 4 - 82894782 - 82900533 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0