Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OTUD6B-AS1 SIRNA Assays ›
          Gene Symbol:
          OTUD6B-AS1
          Gene Name:
          OTUD6B antisense RNA 1 (head to head)
          Gene Aliases:
          GS1-251I9.4
          Chromosome Location:
          Chr.8: 91059906 - 91070189 on Build GRCh38
          UniGene ID:
          Hs.153412
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n511706

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: OTUD6B-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_110438.1 2 398
          NR_110439.1 2 398
          GenBank mRNA BC030623.1 - 2 403
          BC105990.1 - 2 305
          BM715864.1 - 2 332
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA024402.1 - 2 299 Chr. 8 - 91069419 - 91070090 [-] on Build GRCh38
          GenBankmRNA DA260430.1 - 2 289 Chr. 8 - 91069405 - 91070080 [-] on Build GRCh38
          GenBankmRNA BG704702.1 - 2 409 Chr. 8 - 91069444 - 91070189 [-] on Build GRCh38
          GenBankmRNA AB371438.1 - 2 262 Chr. 8 - 91069128 - 91070053 [-] on Build GRCh38
          GenBankmRNA BG528387.1 - 2 331 Chr. 8 - 91063481 - 91070115 [-] on Build GRCh38
          GenBankmRNA BF667646.1 - 248 Chr. - - [-] on Build
          GenBankmRNA DA165632.1 - 2 217 Chr. 8 - 91069331 - 91070008 [-] on Build GRCh38
          GenBankmRNA BM503514.1 - 2 181 Chr. 8 - 91069438 - 91069972 [-] on Build GRCh38
          GenBankmRNA DA829556.1 - 2 294 Chr. 8 - 91069256 - 91070082 [-] on Build GRCh38
          GenBankmRNA BI601380.1 - 2 253 Chr. 8 - 91069206 - 91070043 [-] on Build GRCh38
          GenBankmRNA BF247188.1 - 2 250 Chr. 8 - 91069223 - 91070041 [-] on Build GRCh38
          GenBankmRNA CB118521.1 - 2 286 Chr. 8 - 91069352 - 91070077 [-] on Build GRCh38
          GenBankmRNA BG699467.1 - 296 Chr. - - [-] on Build
          GenBankmRNA DB103066.1 - 2 247 Chr. 8 - 91069387 - 91070039 [-] on Build GRCh38
          GenBankmRNA HY010814.1 - 2 248 Chr. 8 - 91069403 - 91070039 [-] on Build GRCh38
          GenBankmRNA BU569421.1 - 2 326 Chr. 8 - 91069256 - 91070115 [-] on Build GRCh38
          GenBankmRNA DA303246.1 - 2 300 Chr. 8 - 91069394 - 91070091 [-] on Build GRCh38
          GenBankmRNA BE565905.1 - 240 Chr. - - [-] on Build
          GenBankmRNA DA812941.1 - 2 248 Chr. 8 - 91063280 - 91070038 [-] on Build GRCh38
          GenBankmRNA DA039495.1 - 2 239 Chr. 8 - 91069352 - 91070030 [-] on Build GRCh38
          GenBankmRNA BQ632676.1 - 2 313 Chr. 8 - 91069351 - 91070102 [-] on Build GRCh38
          GenBankmRNA BU172165.1 - 2 288 Chr. 8 - 91069195 - 91070079 [-] on Build GRCh38
          GenBankmRNA AV725083.1 - 2 356 Chr. 8 - 91063522 - 91070021 [-] on Build GRCh38
          GenBankmRNA DT220355.1 - 2 323 Chr. 8 - 91069397 - 91070113 [-] on Build GRCh38
          GenBankmRNA DA447987.1 - 2 314 Chr. 8 - 91069436 - 91070105 [-] on Build GRCh38
          GenBankmRNA CB120194.1 - 2 234 Chr. 8 - 91069387 - 91070025 [-] on Build GRCh38
          GenBankmRNA BQ631803.1 - 2 243 Chr. 8 - 91069284 - 91070034 [-] on Build GRCh38
          GenBankmRNA BM546159.1 - 2 248 Chr. 8 - 91068805 - 91070030 [-] on Build GRCh38
          GenBankmRNA BM509004.1 - 2 303 Chr. 8 - 91069400 - 91070087 [-] on Build GRCh38
          GenBankmRNA DB265743.1 - 2 239 Chr. 8 - 91069342 - 91070030 [-] on Build GRCh38
          GenBankmRNA CB132486.1 - 2 229 Chr. 8 - 91069365 - 91070020 [-] on Build GRCh38
          GenBankmRNA BY799587.2 - 2 262 Chr. 8 - 91069407 - 91070053 [-] on Build GRCh38
          GenBankmRNA DB487816.1 - 2 248 Chr. 8 - 91069520 - 91070039 [-] on Build GRCh38
          GenBankmRNA AL532758.3 - 2 249 Chr. 8 - 91068856 - 91070040 [-] on Build GRCh38
          GenBankmRNA DB106765.1 - 2 257 Chr. 8 - 91069356 - 91070048 [-] on Build GRCh38
          GenBankmRNA CD696748.1 - 2 328 Chr. 8 - 91069539 - 91070097 [-] on Build GRCh38
          GenBankmRNA DA094126.1 - 2 232 Chr. 8 - 91063521 - 91070023 [-] on Build GRCh38
          GenBankmRNA DB481667.1 - 2 231 Chr. 8 - 91069434 - 91070022 [-] on Build GRCh38
          GenBankmRNA DB452795.1 - 2 248 Chr. 8 - 91069451 - 91070039 [-] on Build GRCh38
          GenBankmRNA AA057559.1 - 2 228 Chr. 8 - 91069513 - 91070019 [-] on Build GRCh38
          GenBankmRNA DA033391.1 - 2 248 Chr. 8 - 91069358 - 91070039 [-] on Build GRCh38
          GenBankmRNA BF577021.1 - 247 Chr. - - [-] on Build
          GenBankmRNA HY171260.1 - 2 322 Chr. 8 - 91069517 - 91070111 [-] on Build GRCh38
          GenBankmRNA BU570220.1 - 2 265 Chr. 8 - 91069349 - 91070053 [-] on Build GRCh38
          GenBankmRNA BU853047.1 - 2 242 Chr. 8 - 91069221 - 91070033 [-] on Build GRCh38
          GenBankmRNA BI824111.1 - 2 271 Chr. 8 - 91063397 - 91070059 [-] on Build GRCh38
          GenBankmRNA DB173611.1 - 2 197 Chr. 8 - 91069309 - 91069988 [-] on Build GRCh38
          GenBankmRNA CB177477.1 - 2 314 Chr. 8 - 91069519 - 91070102 [-] on Build GRCh38
          GenBankmRNA BE972763.1 - 2 274 Chr. 8 - 91069198 - 91070060 [-] on Build GRCh38
          GenBankmRNA HY146757.1 - 2 263 Chr. 8 - 91069465 - 91070053 [-] on Build GRCh38
          GenBankmRNA DA157027.1 - 2 292 Chr. 8 - 91069418 - 91070080 [-] on Build GRCh38
          GenBankmRNA BP871993.1 - 2 324 Chr. 8 - 91069426 - 91070115 [-] on Build GRCh38
          GenBankmRNA BX502356.1 - 2 249 Chr. 8 - 91069429 - 91070039 [-] on Build GRCh38
          GenBankmRNA DA255024.1 - 2 321 Chr. 8 - 91069456 - 91070111 [-] on Build GRCh38
          GenBankmRNA DN914588.1 - 2 235 Chr. 8 - 91069308 - 91070025 [-] on Build GRCh38
          GenBankmRNA BU569666.1 - 2 252 Chr. 8 - 91063510 - 91070043 [-] on Build GRCh38
          GenBankmRNA AV751156.1 - 2 219 Chr. 8 - 91069424 - 91070010 [-] on Build GRCh38
          GenBankmRNA BM152347.1 - 2 263 Chr. 8 - 91069565 - 91070053 [-] on Build GRCh38
          GenBankmRNA BU188109.1 - 2 265 Chr. 8 - 91069151 - 91070053 [-] on Build GRCh38
          GenBankmRNA DN915989.1 - 2 344 Chr. 8 - 91069512 - 91070109 [-] on Build GRCh38
          GenBankmRNA DA159897.1 - 2 323 Chr. 8 - 91069428 - 91070114 [-] on Build GRCh38
          GenBankmRNA BU952250.1 - 2 238 Chr. 8 - 91069263 - 91070026 [-] on Build GRCh38
          GenBankmRNA BI599069.1 - 2 253 Chr. 8 - 91069231 - 91070043 [-] on Build GRCh38
          GenBankmRNA BU955253.1 - 265 Chr. - - [-] on Build
          GenBankmRNA BJ997080.1 - 2 324 Chr. 8 - 91069568 - 91070115 [-] on Build GRCh38
          GenBankmRNA CD107059.1 - 2 269 Chr. 8 - 91069053 - 91070040 [-] on Build GRCh38
          GenBankmRNA DB447119.1 - 2 248 Chr. 8 - 91069513 - 91070039 [-] on Build GRCh38
          GenBankmRNA BU160824.1 - 2 265 Chr. 8 - 91069175 - 91070053 [-] on Build GRCh38
          GenBankmRNA BM352418.1 - 2 321 Chr. 8 - 91069420 - 91070102 [-] on Build GRCh38
          GenBankmRNA DA156972.1 - 2 228 Chr. 8 - 91069334 - 91070019 [-] on Build GRCh38
          GenBankmRNA DA130457.1 - 2 209 Chr. 8 - 91069334 - 91069999 [-] on Build GRCh38
          GenBankmRNA BU159364.1 - 2 229 Chr. 8 - 91069016 - 91070020 [-] on Build GRCh38
          GenBankmRNA BM905024.1 - 273 Chr. - - [-] on Build
          GenBankmRNA DB472212.1 - 2 248 Chr. 8 - 91069433 - 91070039 [-] on Build GRCh38
          GenBankmRNA T35512.1 - 2 227 Chr. 8 - 91069528 - 91070018 [-] on Build GRCh38
          GenBankmRNA DA833715.1 - 2 347 Chr. 8 - 91069357 - 91070105 [-] on Build GRCh38
          GenBankmRNA DR762734.1 - 2 238 Chr. 8 - 91069078 - 91070029 [-] on Build GRCh38
          GenBankmRNA BI831785.1 - 20 Chr. - - [-] on Build
          GenBankmRNA BG290609.1 - 2 228 Chr. 8 - 91069094 - 91070019 [-] on Build GRCh38
          GenBankmRNA BF574970.1 - 2 248 Chr. 8 - 91069240 - 91070039 [-] on Build GRCh38
          GenBankmRNA DA840239.1 - 2 248 Chr. 8 - 91069385 - 91070039 [-] on Build GRCh38
          GenBankmRNA BI759621.1 - 2 197 Chr. 8 - 91069173 - 91069988 [-] on Build GRCh38
          GenBankmRNA BQ230179.1 - 2 300 Chr. 8 - 91069165 - 91070091 [-] on Build GRCh38
          GenBankmRNA BX957028.1 - 2 250 Chr. 8 - 91069303 - 91070035 [-] on Build GRCh38
          GenBankmRNA DA121039.1 - 2 264 Chr. 8 - 91063571 - 91070054 [-] on Build GRCh38
          GenBankmRNA BF541012.1 - 2 306 Chr. 8 - 91069327 - 91070097 [-] on Build GRCh38
          GenBankmRNA BI545472.1 - 2 253 Chr. 8 - 91069257 - 91070043 [-] on Build GRCh38
          GenBankmRNA BG778864.1 - 2 263 Chr. 8 - 91069247 - 91070053 [-] on Build GRCh38
          GenBankmRNA CN387718.1 - 2 247 Chr. 8 - 91069177 - 91070038 [-] on Build GRCh38
          GenBankmRNA DA317375.1 - 2 321 Chr. 8 - 91063596 - 91070112 [-] on Build GRCh38
          GenBankmRNA CB052237.1 - 2 248 Chr. 8 - 91063691 - 91070032 [-] on Build GRCh38
          GenBankmRNA DA272628.1 - 2 259 Chr. 8 - 91069375 - 91070050 [-] on Build GRCh38
          GenBankmRNA BG502674.1 - 2 266 Chr. 8 - 91069213 - 91070056 [-] on Build GRCh38
          GenBankmRNA BI912732.1 - 2 241 Chr. 8 - 91069140 - 91070033 [-] on Build GRCh38
          GenBankmRNA AV718180.1 - 2 356 Chr. 8 - 91069333 - 91070147 [-] on Build GRCh38
          GenBankmRNA HY047845.1 - 2 248 Chr. 8 - 91063649 - 91070039 [-] on Build GRCh38
          GenBankmRNA BF207734.1 - 2 249 Chr. 8 - 91069214 - 91070039 [-] on Build GRCh38
          GenBankmRNA DB465278.1 - 2 215 Chr. 8 - 91069461 - 91070006 [-] on Build GRCh38
          GenBankmRNA BE738150.1 - 226 Chr. - - [-] on Build
          GenBankmRNA CB107298.1 - 2 209 Chr. 8 - 91063695 - 91070000 [-] on Build GRCh38
          GenBankmRNA BM792763.1 - 1 49 Chr. 8 - 91063519 - 91069732 [-] on Build GRCh38
          GenBankmRNA AL601160.1 - 2 246 Chr. 8 - 91069502 - 91070036 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0