Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OTUD6B-AS1 SIRNA Assays ›
          Gene Symbol:
          OTUD6B-AS1
          Gene Name:
          OTUD6B antisense RNA 1 (head to head)
          Gene Aliases:
          GS1-251I9.4
          Chromosome Location:
          Chr.8: 91059906 - 91070189 on Build GRCh38
          UniGene ID:
          Hs.153412
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n511707

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: OTUD6B-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_110439.1 2 726
          GenBank mRNA BC030623.1 - 2 731
          BC105990.1 - 2 633
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BG290609.1 - 2 557 Chr. 8 - 91069094 - 91070019 [-] on Build GRCh38
          GenBankmRNA BQ631803.1 - 2 571 Chr. 8 - 91069284 - 91070034 [-] on Build GRCh38
          GenBankmRNA AW958749.1 - 3 273 Chr. 8 - 91069013 - 91069995 [-] on Build GRCh38
          GenBankmRNA AB371438.1 - 2 590 Chr. 8 - 91069128 - 91070053 [-] on Build GRCh38
          GenBankmRNA BM546159.1 - 2 576 Chr. 8 - 91068805 - 91070030 [-] on Build GRCh38
          GenBankmRNA DB265743.1 - 2 567 Chr. 8 - 91069342 - 91070030 [-] on Build GRCh38
          GenBankmRNA BQ230179.1 - 2 628 Chr. 8 - 91069165 - 91070091 [-] on Build GRCh38
          GenBankmRNA BI759621.1 - 2 525 Chr. 8 - 91069173 - 91069988 [-] on Build GRCh38
          GenBankmRNA DN914588.1 - 2 563 Chr. 8 - 91069308 - 91070025 [-] on Build GRCh38
          GenBankmRNA BX957028.1 - 2 578 Chr. 8 - 91069303 - 91070035 [-] on Build GRCh38
          GenBankmRNA DA165632.1 - 2 545 Chr. 8 - 91069331 - 91070008 [-] on Build GRCh38
          GenBankmRNA BU188109.1 - 2 593 Chr. 8 - 91069151 - 91070053 [-] on Build GRCh38
          GenBankmRNA DA829556.1 - 2 617 Chr. 8 - 91069256 - 91070082 [-] on Build GRCh38
          GenBankmRNA BI545472.1 - 2 583 Chr. 8 - 91069257 - 91070043 [-] on Build GRCh38
          GenBankmRNA BU952250.1 - 2 566 Chr. 8 - 91069263 - 91070026 [-] on Build GRCh38
          GenBankmRNA AL532758.3 - 2 577 Chr. 8 - 91068856 - 91070040 [-] on Build GRCh38
          GenBankmRNA BI599069.1 - 2 584 Chr. 8 - 91069231 - 91070043 [-] on Build GRCh38
          GenBankmRNA CN387718.1 - 2 575 Chr. 8 - 91069177 - 91070038 [-] on Build GRCh38
          GenBankmRNA BU569421.1 - 2 654 Chr. 8 - 91069256 - 91070115 [-] on Build GRCh38
          GenBankmRNA BG502674.1 - 2 606 Chr. 8 - 91069213 - 91070056 [-] on Build GRCh38
          GenBankmRNA CD107059.1 - 2 597 Chr. 8 - 91069053 - 91070040 [-] on Build GRCh38
          GenBankmRNA BI912732.1 - 2 569 Chr. 8 - 91069140 - 91070033 [-] on Build GRCh38
          GenBankmRNA BU160824.1 - 2 593 Chr. 8 - 91069175 - 91070053 [-] on Build GRCh38
          GenBankmRNA DA156972.1 - 2 556 Chr. 8 - 91069334 - 91070019 [-] on Build GRCh38
          GenBankmRNA DA130457.1 - 2 536 Chr. 8 - 91069334 - 91069999 [-] on Build GRCh38
          GenBankmRNA BU172165.1 - 2 616 Chr. 8 - 91069195 - 91070079 [-] on Build GRCh38
          GenBankmRNA BU159364.1 - 2 557 Chr. 8 - 91069016 - 91070020 [-] on Build GRCh38
          GenBankmRNA BE738150.1 - 555 Chr. - - [-] on Build
          GenBankmRNA DR762734.1 - 2 566 Chr. 8 - 91069078 - 91070029 [-] on Build GRCh38
          GenBankmRNA DB173611.1 - 2 525 Chr. 8 - 91069309 - 91069988 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0