Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR762HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110940.1 | 3 | 1386 | |||
NR_110941.1 | 2 | 1141 | ||||
NR_110942.1 | 3 | 598 | ||||
GenBank mRNA | BM769724.1 | - | 3 | 597 |